accessibility menu, dialog, popup

UserName

CAS RN 699-02-5

CH3OH

CAS RN 699-02-5

IUPAC Name: 2-(4-methylphenyl)ethan-1-ol
Synonyms: 2-P-cresylethanol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: DAVFJRVIVZOKKS-UHFFFAOYSA-N
SMILES: CC1=CC=C(CCO)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results