accessibility menu, dialog, popup

UserName

CAS RN 69824-23-3

IUPAC Name: 2-(2-methylpropyl)-1,3-dithiane
Synonyms: 2-(2-methylpropyl)-1,3-dithiane
Molecular Weight (g/mol): 176.34
Molecular Formula: C8H16S2
InChi Key: WGIHYSKTNRQNHP-UHFFFAOYSA-N
SMILES: CC(C)CC1SCCCS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results