accessibility menu, dialog, popup

UserName

CAS RN 69807-81-4

CH3NH2N

CAS RN 69807-81-4

IUPAC Name: 1-(1-methyl-1H-pyrrol-2-yl)methanamine
Synonyms: 1-(1-methylpyrrol-2-yl)methanamine
Molecular Weight (g/mol): 110.16
Molecular Formula: C6H10N2
InChi Key: GGCBARJYVAPZJQ-UHFFFAOYSA-N
SMILES: CN1C=CC=C1CN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)