accessibility menu, dialog, popup

UserName

CH3OOOOH

CAS RN 69716-05-8

IUPAC Name: 2-(6-methoxy-1-benzofuran-3-yl)acetic acid
Synonyms: (6-methoxy-1-benzofuran-3-yl)acetic acid
Molecular Weight (g/mol): 206.2
Molecular Formula: C11H10O4
InChi Key: QCXJFLREQGIACT-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(CC(O)=O)=COC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results