accessibility menu, dialog, popup

UserName

CAS RN 697-73-4

FFCl

CAS RN 697-73-4

IUPAC Name: 2-(chloromethyl)-1,3-difluorobenzene
Synonyms: 2-(chloromethyl)-1,3-difluorobenzene
Molecular Weight (g/mol): 162.56
Molecular Formula: C7H5ClF2
InChi Key: MJXRENZUAQXZGJ-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1CCl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results