accessibility menu, dialog, popup

UserName

ClSClOO

CAS RN 6966-45-6

IUPAC Name: (4-chlorophenyl)methanesulfonyl chloride
Synonyms: (4-chlorophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 225.08
Molecular Formula: C7H6Cl2O2S
InChi Key: DBJRPJSDYFDWPV-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CS(Cl)(=O)=O)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results