accessibility menu, dialog, popup

UserName

CH3OONBrNH2N

CAS RN 6966-01-4

IUPAC Name: methyl 3-amino-6-bromopyrazine-2-carboxylate
Synonyms: methyl 3-amino-6-bromopyrazine-2-carboxylate
Molecular Weight (g/mol): 232.04
Molecular Formula: C6H6BrN3O2
InChi Key: CNXSIRHOIFRMOB-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC(Br)=CN=C1N
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results