accessibility menu, dialog, popup

UserName

HClClN

CAS RN 6959-47-3

IUPAC Name: hydrogen 2-(chloromethyl)pyridine chloride
Synonyms: hydrogen 2-(chloromethyl)pyridine chloride
Molecular Weight (g/mol): 164.03
Molecular Formula: C6H7Cl2N
InChi Key: JPMRGPPMXHGKRO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClCC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)