CAS RN 6943-17-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Nitro-4-quinazolone, TRC
CAS: 6943-17-5 Molecular Formula: C8H5N3O3 Molecular Weight (g/mol): 191.14 Synonym: 6-Nitro-3H-quinazolin-4-one,6-Nitro-4(1H)-quinazolinone,6-Nitroquinazolin-4-ol,6-Nitro-3,4-dihydroquinazolin-4-one,NSC 51669 IUPAC Name: 6-nitro-1H-quinazolin-4-one SMILES: [O-][N+](=O)c1ccc2NC=NC(=O)c2c1