accessibility menu, dialog, popup

UserName

OHOHOBr

CAS RN 6940-50-7

IUPAC Name: 2-(4-bromophenyl)-2-hydroxyacetic acid
Synonyms: p-bromomandelic acid
Molecular Weight (g/mol): 231.05
Molecular Formula: C8H7BrO3
InChi Key: BHZBRPQOYFDTAB-UHFFFAOYNA-N
SMILES: OC(C(O)=O)C1=CC=C(Br)C=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results