accessibility menu, dialog, popup

UserName

CAS RN 694-28-0

ClO

CAS RN 694-28-0

IUPAC Name: 2-chlorocyclopentan-1-one
Synonyms: cyclopentanone, 2-chloro-
Molecular Weight (g/mol): 118.56
Molecular Formula: C5H7ClO
InChi Key: AXDZFGRFZOQVBV-UHFFFAOYNA-N
SMILES: ClC1CCCC1=O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)