CAS RN 6939-95-3
IUPAC Name:
1-(3-chlorophenyl)ethan-1-ol
Synonyms:
1-(3-chlorophenyl)ethanol
Molecular Weight (g/mol):
156.61
Molecular Formula:
C8H9ClO
InChi Key:
QYUQVBHGBPRDKN-UHFFFAOYNA-N
SMILES:
CC(O)C1=CC=CC(Cl)=C1
1
–
1
of
1
results
1
1-(3-Chlorophenyl)ethanol, 97%
CAS: 6939-95-3 Molecular Formula: C8H9ClO Molecular Weight (g/mol): 156.609 MDL Number: MFCD00021864 InChI Key: QYUQVBHGBPRDKN-UHFFFAOYSA-N Synonym: 1-3-chlorophenyl ethanol,3-chloro-alpha-methylbenzyl alcohol,1-3-chlorophenyl ethan-1-ol,1-3-chlorophenyl-1-ethanol,3-chlorophenylethanol,acmc-1cap8,1-m-chlorophenyl ethanol,3'-chloro-1-phenylethanol,1-3-chlorophenyl-ethanol,1-3-chlorophenyl ethyl alcohol PubChem CID: 97779 IUPAC Name: 1-(3-chlorophenyl)ethanol SMILES: CC(C1=CC(=CC=C1)Cl)O