accessibility menu, dialog, popup

UserName

CH3NH2Br

CAS RN 6933-10-4

IUPAC Name: 4-bromo-3-methylaniline
Synonyms: 3-methyl-4-bromoaniline
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: MMEGELSFOYDPQW-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC=C1Br
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results