accessibility menu, dialog, popup

UserName

HOHNO

CAS RN 693-06-1

IUPAC Name: N-(2-hydroxyethyl)formamide
Synonyms: N-(2-hydroxyethyl)formamide
Molecular Weight (g/mol): 89.09
Molecular Formula: C3H7NO2
InChi Key: BAMUPQJDKBGDPU-UHFFFAOYSA-N
SMILES: OCCNC=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results