accessibility menu, dialog, popup

UserName

H3CO(R)O(R)HO(S)O(R)O(R)HO(S)OH(S)OH(R)OH(R)OH(R)OH

CAS RN 69227-93-6

IUPAC Name: (2R,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6R)-6-(dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: (2R,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6R)-6-(dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 510.62
Molecular Formula: C24H46O11
InChi Key: NLEBIOOXCVAHBD-QKMCSOCLSA-N
SMILES: CCCCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O
Quantity 
  • (5)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (6)
  • (3)