accessibility menu, dialog, popup

UserName

H3C(S)OHOH

CAS RN 6920-22-5

IUPAC Name: (2S)-hexane-1,2-diol
Synonyms: 1,2-hexanediol
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: FHKSXSQHXQEMOK-LURJTMIESA-N
SMILES: CCCC[C@H](O)CO
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)