accessibility menu, dialog, popup

UserName

CAS RN 690632-72-5

BOBrHOOH

CAS RN 690632-72-5

IUPAC Name: (5-bromo-2,3-dihydro-1-benzofuran-7-yl)boronic acid
Synonyms: 5-bromo-2,3-dihydro-1-benzofuran-7-ylboronic acid
Molecular Weight (g/mol): 242.86
Molecular Formula: C8H8BBrO3
InChi Key: BSSWVOKZPQTKGW-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Br)=CC2=C1OCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results