accessibility menu, dialog, popup

UserName

CAS RN 690632-39-4

NNO

CAS RN 690632-39-4

IUPAC Name: 2-(pyrrolidin-1-yl)pyridine-3-carbaldehyde
Synonyms: 2-(pyrrolidin-1-yl)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 176.22
Molecular Formula: C10H12N2O
InChi Key: FTACFSVJFQMXQE-UHFFFAOYSA-N
SMILES: O=CC1=C(N=CC=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results