accessibility menu, dialog, popup

UserName

FFFNOHNH2(E/Z)

CAS RN 690632-34-9

IUPAC Name: 2,4,5-trifluoro-N'-hydroxybenzene-1-carboximidamide
Synonyms: 2,4,5-trifluoro-N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol): 190.13
Molecular Formula: C7H5F3N2O
InChi Key: JWJFTTGUXZRMMI-UHFFFAOYSA-N
SMILES: NC(=NO)C1=CC(F)=C(F)C=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results