accessibility menu, dialog, popup

UserName

NNHO

CAS RN 690631-99-3

IUPAC Name: [6-(piperidin-1-yl)pyridin-3-yl]methanol
Synonyms: [6-(piperidin-1-yl)pyridin-3-yl]methanol
Molecular Weight (g/mol): 192.26
Molecular Formula: C11H16N2O
InChi Key: VLAMDMADWWGEDH-UHFFFAOYSA-N
SMILES: OCC1=CN=C(C=C1)N1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results