accessibility menu, dialog, popup

UserName

CAS RN 687-47-8

H3COO(S)CH3OH

CAS RN 687-47-8

IUPAC Name: ethyl (2S)-2-hydroxypropanoate
Synonyms: ethyl lactate
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: LZCLXQDLBQLTDK-BYPYZUCNSA-N
SMILES: CCOC(=O)[C@H](C)O
Molecular Weight (g/mol) 
  • (4)
  • (5)
Boiling Point 
  • (3)
  • (5)
Percent Purity 
  • (1)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (3)
  • (4)
  • (5)
  • (3)
  • (5)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)