accessibility menu, dialog, popup

UserName

CAS RN 68694-11-1

H3CONClFFFNN(E/Z)

CAS RN 68694-11-1

IUPAC Name: N-[4-chloro-2-(trifluoromethyl)phenyl]-1-(1H-imidazol-1-yl)-2-propoxyethan-1-imine
Synonyms: N-[4-chloro-2-(trifluoromethyl)phenyl]-1-(imidazol-1-yl)-2-propoxyethanimine
Molecular Weight (g/mol): 345.75
Molecular Formula: C15H15ClF3N3O
InChi Key: HSMVPDGQOIQYSR-UHFFFAOYSA-N
SMILES: CCCOCC(=NC1=CC=C(Cl)C=C1C(F)(F)F)N1C=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results