CAS RN 68592-11-0
CAS RN 68592-11-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro-5-(1-chloro-1,3-dihydro-3-oxo-1-isobenzofuranyl)-benzenesulfonyl Chloride, TRC
CAS: 68592-11-0 Molecular Formula: C14H7Cl3O4S Molecular Weight (g/mol): 377.63 Synonym: 3-Chloro-3-[4-chloro-3-(chlorosulfonyl)phenyl]-phthalide IUPAC Name: 2-chloro-5-(1-chloro-3-oxo-2-benzofuran-1-yl)benzenesulfonyl chloride SMILES: O=C1OC(Cl)(C2=CC=C(Cl)C(=C2)S(=O)(=O)Cl)C=3C=CC=CC13