accessibility menu, dialog, popup

UserName

CAS RN 68528-80-3

IUPAC Name: bis(2,5-dioxopyrrolidin-1-yl) octanedioate
Synonyms: disuccinimidyl suberate
Molecular Weight (g/mol): 368.34
Molecular Formula: C16H20N2O8
InChi Key: ZWIBGKZDAWNIFC-UHFFFAOYSA-N
SMILES: O=C(CCCCCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)