CAS RN 68504-35-8
IUPAC Name:
tetrasodium (3Z,6Z)-4,5-dioxo-3,6-bis[2-(2-sulfonatophenyl)hydrazin-1-ylidene]-3,4,5,6-tetrahydronaphthalene-2,7-disulfonate
Synonyms:
tetrasodium (3Z,6Z)-4,5-dioxo-3,6-bis[2-(2-sulfonatophenyl)hydrazin-1-ylidene]naphthalene-2,7-disulfonate
Molecular Weight (g/mol):
776.55
Molecular Formula:
C22H12N4Na4O14S4
InChi Key:
OWVXGCUJHKOBMQ-DTSPTRIMSA-J
SMILES:
[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1N\N=C1\C(=O)C2=C(C=C1S([O-])(=O)=O)C=C(\C(=N/NC1=CC=CC=C1S([O-])(=O)=O)C2=O)S([O-])(=O)=O
1
–
1
of
1
results
1
Sulfonazo III salt
CAS: 68504-35-8 Molecular Formula: C22H16N4Na4O14S4 Molecular Weight (g/mol): 780.583 MDL Number: MFCD00003942 InChI Key: WHVMUCGHHVLOCE-FMNKGCAFSA-N Synonym: Tetrasodium 4,5-dihydroxy-3,6-bis[(2-sulfonatophenyl)azo]naphthalene-2,7-disulfonate PubChem CID: 131857569 IUPAC Name: (3E,6E)-4,5-dioxo-3,6-bis[(2-sulfophenyl)hydrazinylidene]naphthalene-2,7-disulfonic acid;sodium SMILES: C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4S(=O)(=O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na].[Na].[Na]