accessibility menu, dialog, popup

UserName

CAS RN 68498-54-4

CN

CAS RN 68498-54-4

IUPAC Name: isocyanocyclopentane
Synonyms: isocyanocyclopentane
Molecular Weight (g/mol): 95.15
Molecular Formula: C6H9N
InChi Key: XTLSDEJNOQVCHL-UHFFFAOYSA-N
SMILES: [C-]#[N+]C1CCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results