accessibility menu, dialog, popup

UserName

CH3OPOOH3CH2C

CAS RN 682-30-4

IUPAC Name: diethyl ethenylphosphonate
Synonyms: diethyl ethenylphosphonate
Molecular Weight (g/mol): 164.14
Molecular Formula: C6H13O3P
InChi Key: DREPONDJUKIQLX-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)C=C
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results