accessibility menu, dialog, popup

UserName

CAS RN 68162-47-0

BBrHOOH

CAS RN 68162-47-0

IUPAC Name: [4-(bromomethyl)phenyl]boronic acid
Synonyms: 4-(bromomethyl)phenylboronic acid
Molecular Weight (g/mol): 214.85
Molecular Formula: C7H8BBrO2
InChi Key: PDNOURKEZJZJNZ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(CBr)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)