accessibility menu, dialog, popup

UserName

CAS RN 68131-04-4

IUPAC Name: disodium bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
Synonyms: sodium humate
Molecular Weight (g/mol): 226.14
Molecular Formula: C9H8Na2O4
InChi Key: HMGUIQPKFUZDPV-UHFFFAOYNA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1C2CC(C=C2)C1C([O-])=O
Grade 
  • (5)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)