accessibility menu, dialog, popup

UserName

OONNOO

CAS RN 6813-38-3

IUPAC Name: [2,2'-bipyridine]-4,4'-dicarboxylate
Synonyms: [2,2'-bipyridine]-4,4'-dicarboxylate
Molecular Weight (g/mol): 242.19
Molecular Formula: C12H6N2O4
InChi Key: FXPLCAKVOYHAJA-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results