CAS RN 68119-31-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)acetonitrile, TRC
CAS: 68119-31-3 Molecular Formula: C9H5NO2F2 Molecular Weight (g/mol): 197.14 SMILES: FC1(F)Oc2ccc(CC#N)cc2O1