accessibility menu, dialog, popup

UserName

OH(S)NHO

CAS RN 68108-18-9

IUPAC Name: (4S)-4-hydroxypyrrolidin-2-one
Synonyms: (4S)-4-hydroxypyrrolidin-2-one
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: IOGISYQVOGVIEU-VKHMYHEASA-N
SMILES: O[C@@H]1CNC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results