accessibility menu, dialog, popup

UserName

CAS RN 681-84-5

CH3OSiOH3COH3COH3C

CAS RN 681-84-5

IUPAC Name: tetramethyl silicate
Synonyms: tetramethyl orthosilicate silicic acid (H4siO4), tetramethyl ester
Molecular Weight (g/mol): 152.22
Molecular Formula: C4H12O4Si
InChi Key: LFQCEHFDDXELDD-UHFFFAOYSA-N
SMILES: CO[Si](OC)(OC)OC
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (4)