accessibility menu, dialog, popup

UserName

ONNHNHN

CAS RN 68-94-0

IUPAC Name: 6,7-dihydro-3H-purin-6-one
Synonyms: hypoxanthine
Molecular Weight (g/mol): 136.11
Molecular Formula: C5H4N4O
InChi Key: FDGQSTZJBFJUBT-UHFFFAOYSA-N
SMILES: O=C1N=CNC2=C1NC=N2
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)