accessibility menu, dialog, popup

UserName

OHOHS

CAS RN 68-11-1

IUPAC Name: 2-sulfanylacetic acid
Synonyms: thioglycolic acid thioglykolsaeure acide thioglycolique kyselina thioglykolova thiovanic acid kyselina merkaptooctova ammonium thioglycolate (product) sodium thioglycolate (substance) thioglycolic acid and its salts ammonium thioglycolate (substance)
Molecular Weight (g/mol): 92.11
Molecular Formula: C2H4O2S
InChi Key: CWERGRDVMFNCDR-UHFFFAOYSA-N
SMILES: OC(=O)CS
Boiling Point 
  • (6)
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (10)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (3)
  • (6)
  • (2)
  • (4)
  • (4)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (10)
  • (3)
  • (4)
  • (2)