accessibility menu, dialog, popup

UserName

CAS RN 6795-87-5

CH3H3COH3C

CAS RN 6795-87-5

IUPAC Name: 2-methoxybutane
Synonyms: sec-butyl methyl ether
Molecular Weight (g/mol): 88.15
Molecular Formula: C5H12O
InChi Key: FVNIMHIOIXPIQT-UHFFFAOYNA-N
SMILES: CCC(C)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results