accessibility menu, dialog, popup

UserName

CAS RN 67858-47-3

CH3NOOH3CO

CAS RN 67858-47-3

IUPAC Name: methyl 4-formyl-1-methyl-1H-pyrrole-2-carboxylate
Synonyms: methyl 4-formyl-1-methylpyrrole-2-carboxylate
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: GXSZDESAIZEFGZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(C=O)=CN1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results