accessibility menu, dialog, popup

UserName

CAS RN 67686-01-5

HON

CAS RN 67686-01-5

IUPAC Name: (1-benzylpiperidin-4-yl)methanol
Synonyms: (1-benzylpiperidin-4-yl)methanol
Molecular Weight (g/mol): 205.3
Molecular Formula: C13H19NO
InChi Key: FLQPYEOKVZYXRL-UHFFFAOYSA-N
SMILES: OCC1CCN(CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results