accessibility menu, dialog, popup

UserName

CAS RN 6759-78-0

OHNN

CAS RN 6759-78-0

IUPAC Name: 8-hydroxyquinoline-2-carbonitrile
Synonyms: 8-hydroxyquinoline-2-carbonitrile
Molecular Weight (g/mol): 170.17
Molecular Formula: C10H6N2O
InChi Key: KUQKKIBQVSFDHX-UHFFFAOYSA-N
SMILES: OC1=C2N=C(C=CC2=CC=C1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)