accessibility menu, dialog, popup

UserName

CAS RN 67478-48-2

IUPAC Name: 2-chloro-1-(triphenylmethyl)-1H-imidazole
Synonyms: 2-chloro-1-(triphenylmethyl)imidazole
Molecular Weight (g/mol): 344.84
Molecular Formula: C22H17ClN2
InChi Key: JTMBWVKMAGHXKI-UHFFFAOYSA-N
SMILES: ClC1=NC=CN1C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results