accessibility menu, dialog, popup

UserName

CAS RN 67387-76-2

CH3OOO

CAS RN 67387-76-2

IUPAC Name: 3-(cyclopentyloxy)-4-methoxybenzaldehyde
Synonyms: 3-(cyclopentyloxy)-4-methoxybenzaldehyde
Molecular Weight (g/mol): 220.27
Molecular Formula: C13H16O3
InChi Key: FZFWPURYSWKIRT-UHFFFAOYSA-N
SMILES: COC1=C(OC2CCCC2)C=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results