CAS RN 6734-33-4
CAS RN 6734-33-4
IUPAC Name:
4-methyl-7-{[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy}-2H-chromen-2-one
Synonyms:
4-methyl-7-{[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy}chromen-2-one
Molecular Weight (g/mol):
308.29
Molecular Formula:
C15H16O7
InChi Key:
JWIYLOHVJDJZOQ-KAOXEZKKSA-N
SMILES:
CC1=CC(=O)OC2=CC(O[C@@H]3OC[C@@H](O)[C@H](O)[C@H]3O)=CC=C12
1
–
4
of
4
results
2
4-Methylumbelliferyl b-D-Xylopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Methylumbelliferyl b-D-Xylopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
4-Methylumbelliferyl b-D-Xylopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.