accessibility menu, dialog, popup

UserName

CAS RN 67305-24-2

CH3ONClO

CAS RN 67305-24-2

IUPAC Name: 5-methyl-1,2-oxazole-4-carbonyl chloride
Synonyms: 5-methyl-1,2-oxazole-4-carbonyl chloride
Molecular Weight (g/mol): 145.54
Molecular Formula: C5H4ClNO2
InChi Key: ZKAQPVQEYCFRTK-UHFFFAOYSA-N
SMILES: CC1=C(C=NO1)C(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results