accessibility menu, dialog, popup

UserName

NH2(R)OHO

CAS RN 673-06-3

IUPAC Name: (2R)-2-amino-3-phenylpropanoic acid
Synonyms: D-phenylalanine phenylalanine D-form
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: COLNVLDHVKWLRT-MRVPVSSYSA-N
SMILES: N[C@H](CC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (4)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)