accessibility menu, dialog, popup

UserName

CAS RN 6705-03-9

IUPAC Name: 2-amino-5-methoxybenzoic acid
Synonyms: 2-amino-5-methoxybenzoic acid
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: UMKSAURFQFUULT-UHFFFAOYSA-N
SMILES: COC1=CC=C(N)C(=C1)C(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results