CAS RN 67032-77-3
CAS RN 67032-77-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Magnesium 2,4-pentanedionate hydrate, typically 98+% (metals basis)
CAS: 67032-77-3 Molecular Formula: C10H14MgO4 Molecular Weight (g/mol): 222.523 MDL Number: MFCD00150168 InChI Key: AKTIAGQCYPCKFX-SYWGCQIGSA-L Synonym: 4-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one PubChem CID: 25113476 IUPAC Name: magnesium;(E)-4-oxopent-2-en-2-olate SMILES: CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Mg+2]