accessibility menu, dialog, popup

UserName

CAS RN 668-94-0

NHN

CAS RN 668-94-0

IUPAC Name: 4,5-diphenyl-1H-imidazole
Synonyms: diphenylimidazole
Molecular Weight (g/mol): 220.28
Molecular Formula: C15H12N2
InChi Key: CPHGOBGXZQKCKI-UHFFFAOYSA-N
SMILES: N1C=NC(=C1C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results