accessibility menu, dialog, popup

UserName

NN

CAS RN 6674-22-2

IUPAC Name: 2H,3H,4H,6H,7H,8H,9H,10H-pyrimido[1,2-a]azepine
Synonyms: 1,8-diazabicycloundec-7-ene
Molecular Weight (g/mol): 152.24
Molecular Formula: C9H16N2
InChi Key: GQHTUMJGOHRCHB-UHFFFAOYSA-N
SMILES: C1CN=C2CCCCCN2C1
Molecular Weight (g/mol) 
  • (6)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (3)
  • (6)
  • (4)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (4)