accessibility menu, dialog, popup

UserName

CAS RN 6665-83-4

HOOO

CAS RN 6665-83-4

IUPAC Name: 6-hydroxy-2-phenyl-4H-chromen-4-one
Synonyms: 6-hydroxyflavone
Molecular Weight (g/mol): 238.24
Molecular Formula: C15H10O3
InChi Key: GPZYYYGYCRFPBU-UHFFFAOYSA-N
SMILES: OC1=CC=C2OC(=CC(=O)C2=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results