accessibility menu, dialog, popup

UserName

OOOHOH

CAS RN 6665-66-3

IUPAC Name: 5,6-dihydroxy-2-phenyl-4H-chromen-4-one
Synonyms: 5,6-dihydroxy-2-phenylchromen-4-one
Molecular Weight (g/mol): 254.24
Molecular Formula: C15H10O4
InChi Key: AGZAGADSYIYYCT-UHFFFAOYSA-N
SMILES: OC1=CC=C2OC(=CC(=O)C2=C1O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results